Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174654.2 | NP_777079.1 | 12 - 13 | 1831 | 105 | |
GenBank mRNA | AJ508718.1 | - | 12 - 13 | 1831 | 105 | |
AY052775.1 | - | 13 - 14 | 1915 | 105 | ||
BC133437.1 | - | 12 - 13 | 1876 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |