Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175716.3 | NP_783647.1 | 1 - 2 | 100 | 91 | |
GenBank mRNA | AF354057.1 | - | 1 - 2 | 102 | 91 | |
KU641032.1 | - | 1 - 2 | 62 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |