Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175718.2 | NP_783649.1 | 1 - 2 | 152 | 59 | |
GenBank mRNA | AJ510148.1 | - | 1 - 2 | 152 | 59 | |
BC102976.1 | - | 1 - 2 | 173 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |