Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175788.1 | NP_786982.1 | 7 - 8 | 1339 | 138 | |
GenBank mRNA | Y11045.1 | - | 1371 | 138 | ||
Y11046.1 | - | 1364 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |