Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176633.3 | NP_788806.1 | 1 - 2 | 858 | 64 | |
GenBank mRNA | AB003485.1 | - | 1 - 2 | 860 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |