Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176640.2 | NP_788813.1 | 1 - 2 | 99 | 118 | |
GenBank mRNA | AJ440723.1 | - | 1 - 2 | 106 | 118 | |
BT021691.1 | - | 1 - 2 | 106 | 118 | ||
BT021802.1 | - | 1 - 2 | 99 | 118 | ||
BT026188.1 | - | 1 - 2 | 90 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |