Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181017.2 | NP_851360.1 | 5 - 6 | 377 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |