Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181667.1 | NP_858053.1 | 6 - 7 | 2140 | 82 | |
GenBank mRNA | AF516915.1 | - | 6 - 7 | 2140 | 82 | |
BC148059.1 | - | 6 - 7 | 2152 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |