Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015649.2 | NP_001015649.1 | 6 - 7 | 837 | 129 | |
GenBank mRNA | BC108139.1 | - | 6 - 7 | 837 | 129 | |
BT020743.1 | - | 6 - 7 | 810 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |