Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033626.1 | NP_001028798.1 | 3 - 4 | 429 | 125 | |
GenBank mRNA | BC102564.1 | - | 3 - 4 | 429 | 125 | |
BC142072.1 | - | 3 - 4 | 427 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |