Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034039.2 | NP_001029211.1 | 19 - 20 | 1415 | 139 | |
GenBank mRNA | BC105184.1 | - | 19 - 20 | 1416 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |