Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034263.1 | NP_001029435.1 | 2 - 3 | 132 | 106 | |
GenBank mRNA | BC102531.1 | - | 2 - 3 | 132 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |