Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034311.2 | NP_001029483.1 | 3 - 4 | 384 | 102 | |
GenBank mRNA | BC103413.1 | - | 3 - 4 | 385 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |