Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034315.1 | NP_001029487.1 | 4 - 5 | 536 | 73 | |
GenBank mRNA | BC105194.1 | - | 4 - 5 | 536 | 73 | |
BT030537.1 | - | 4 - 5 | 524 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |