Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034323.2 | NP_001029495.1 | 1 - 2 | 99 | 51 | |
GenBank mRNA | BC102721.1 | - | 1 - 2 | 101 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |