Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034345.1 | NP_001029517.1 | 2 - 3 | 319 | 131 | |
GenBank mRNA | BC103203.1 | - | 2 - 3 | 319 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |