Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM86B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034528.1 | NP_001029700.1 | 1 - 2 | 67 | 77 | |
GenBank mRNA | BC102239.1 | - | 1 - 2 | 67 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |