Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034618.2 | NP_001029790.1 | 17 - 18 | 2432 | 65 | |
GenBank mRNA | BC103352.1 | - | 17 - 18 | 2433 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |