Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034628.3 | NP_001029800.1 | 6 - 7 | 1131 | 85 | |
NM_001320319.1 | NP_001307248.1 | 6 - 7 | 1131 | 85 | ||
NR_135204.1 | 5 - 6 | 955 | 85 | |||
GenBank mRNA | BC102133.1 | - | 6 - 7 | 727 | 85 | |
CO872328.1 | - | 1 - 2 | 54 | 85 | ||
DV882028.1 | - | 6 - 7 | 712 | 85 | ||
EH162161.1 | - | 5 - 6 | 577 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |