Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034649.2 | NP_001029821.1 | 5 - 6 | 884 | 120 | |
GenBank mRNA | BC102468.1 | - | 5 - 6 | 885 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |