Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034673.2 | NP_001029845.1 | 2 - 3 | 244 | 108 | |
GenBank mRNA | BC103369.1 | - | 2 - 3 | 246 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |