Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD3B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034696.1 | NP_001029868.1 | 5 - 6 | 613 | 88 | |
GenBank mRNA | BC105259.1 | - | 5 - 6 | 613 | 88 | |
BT030700.1 | - | 5 - 6 | 614 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |