Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035021.1 | NP_001030193.1 | 4 - 5 | 598 | 68 | |
GenBank mRNA | BC103359.1 | - | 4 - 5 | 598 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |