Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035064.1 | NP_001030236.1 | 8 - 9 | 766 | 123 | |
GenBank mRNA | BC104588.1 | - | 8 - 9 | 766 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |