Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035302.2 | NP_001030379.1 | 1 - 2 | 221 | 114 | |
GenBank mRNA | BC102679.1 | - | 1 - 2 | 222 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |