Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035407.2 | NP_001030484.2 | 3 - 4 | 423 | 67 | |
NM_001166367.1 | NP_001159839.1 | 3 - 4 | 423 | 67 | ||
GenBank mRNA | BC102142.1 | - | 2 - 3 | 196 | 67 | |
BC148158.1 | - | 2 - 3 | 235 | 67 | ||
EV623524.1 | - | 3 - 4 | 423 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |