Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040563.1 | NP_001035653.1 | 5 - 6 | 619 | 90 | |
GenBank mRNA | BC104577.1 | - | 5 - 6 | 619 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |