Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORCS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040565.1 | NP_001035655.2 | 1 | 1749 | 115 | |
GenBank mRNA | BC104551.1 | - | 1 | 1749 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |