Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5F1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075136.2 | NP_001068604.1 | 1 - 2 | 86 | 113 | |
GenBank mRNA | BC102466.1 | - | 1 - 2 | 87 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |