Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075900.1 | NP_001069368.1 | 4 - 5 | 660 | 79 | |
GenBank mRNA | BC102549.1 | - | 4 - 5 | 660 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |