Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076467.1 | NP_001069935.1 | 5 - 6 | 463 | 131 | |
NM_001370742.1 | NP_001357671.1 | 5 - 6 | 421 | 131 | ||
GenBank mRNA | BC102743.1 | - | 5 - 6 | 463 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |