Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076998.2 | NP_001070466.1 | 5 - 6 | 370 | 79 | |
GenBank mRNA | BC103039.1 | - | 5 - 6 | 371 | 79 | |
BC105558.1 | - | 3 - 4 | 1156 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |