Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174345.4 | NP_776770.2 | 6 - 7 | 1392 | 66 | |
GenBank mRNA | BC105182.1 | - | 6 - 7 | 1393 | 66 | |
BC154389.1 | - | 2 - 3 | 510 | 66 | ||
BT030487.1 | - | 6 - 7 | 1378 | 66 | ||
X53335.1 | - | 1317 | 66 | |||
X53827.1 | - | 1351 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |