Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174731.4 | NP_777156.1 | 1 - 2 | 106 | 61 | |
GenBank mRNA | AY911375.1 | - | 1 - 2 | 56 | 61 | |
BC102873.1 | - | 1 - 2 | 107 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |