Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024509.3 | NP_001019680.2 | 8 - 9 | 638 | 88 | |
GenBank mRNA | BC105379.1 | - | 8 - 9 | 639 | 88 | |
DQ010410.1 | - | 8 - 9 | 556 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |