Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC109956.1 | - | 7 - 8 | 997 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |