Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037616.1 | NP_001032705.1 | 2 - 3 | 275 | 82 | |
GenBank mRNA | BC109755.1 | - | 2 - 3 | 275 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |