Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038017.2 | NP_001033106.1 | 6 - 7 | 983 | 123 | |
GenBank mRNA | AY533858.1 | - | 2 - 3 | 182 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |