Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038093.2 | NP_001033182.1 | 1 - 2 | 128 | 130 | |
GenBank mRNA | BC111280.1 | - | 1 - 2 | 130 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |