Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038102.2 | NP_001033191.2 | 1 - 2 | 302 | 123 | |
GenBank mRNA | BC111246.1 | - | 1 - 2 | 256 | 123 | |
EV636615.1 | - | 1 - 2 | 306 | 123 |
Gene Symbol: PIGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270946.1 | NP_001257875.1 | 1 - 2 | 302 | 123 | |
GenBank mRNA | EH163221.1 | - | 1 - 2 | 265 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Inventoried Probe spans exons |