Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038221.2 | NP_001033310.1 | 11 - 12 | 1404 | 141 | |
GenBank mRNA | BC111193.1 | - | 11 - 12 | 1405 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |