Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038528.2 | NP_001033617.1 | 1 - 2 | 53 | 79 | |
GenBank mRNA | BC109990.1 | - | 1 - 2 | 54 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |