Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038531.2 | NP_001033620.1 | 7 - 8 | 898 | 64 | |
GenBank mRNA | BC109865.1 | - | 7 - 8 | 899 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |