Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038534.2 | NP_001033623.1 | 9 - 10 | 1424 | 70 | |
GenBank mRNA | BC109682.1 | - | 9 - 10 | 1425 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |