Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038544.2 | NP_001033633.1 | 2 - 3 | 370 | 66 | |
GenBank mRNA | BC109825.1 | - | 2 - 3 | 371 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |