Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038544.2 | NP_001033633.1 | 5 - 6 | 606 | 113 | |
GenBank mRNA | BC109825.1 | - | 5 - 6 | 607 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |