Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038545.2 | NP_001033634.1 | 2 - 3 | 218 | 145 | |
GenBank mRNA | BC109977.1 | - | 2 - 3 | 219 | 145 | |
BT030697.1 | - | 2 - 3 | 188 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |