Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038549.1 | NP_001033638.1 | 12 - 13 | 1468 | 142 | |
GenBank mRNA | BC109786.1 | - | 12 - 13 | 1468 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |