Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038634.1 | NP_001033723.1 | 6 - 7 | 735 | 83 | |
GenBank mRNA | BC148924.1 | - | 6 - 7 | 741 | 83 | |
BT029862.1 | - | 6 - 7 | 635 | 83 | ||
DQ116783.1 | - | 6 - 7 | 735 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |