Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038680.1 | NP_001033769.1 | 5 - 6 | 820 | 71 | |
GenBank mRNA | BC110129.1 | - | 5 - 6 | 820 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |